EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Nephroprotective effect of vanillic acid against cisplatin induced nephrotoxicity in wistar rats: a biochemical and molecular study.
PMID 25561092
Abstract
Cisplatin is one of the extensively used anticancer drugs against various cancers. Dosage dependent nephrotoxicity is the major problem in cisplatin chemotherapy. Cisplatin induced nephrotoxicity results in the depletion of renal antioxidant defence system. Our present study is aimed to investigate the nephroprotective effect of vanilic acid to against cisplatin induced nephrotoxicity in male wistar rats. Elevated levels of serum creatinine, blood urea nitrogen, serum uric acid and reduced antioxidant status were observed as indicatives of nephrotoxicity in cisplatin (7mg/kg bw) alone administered rats. Animals which are pre-treated with vanillic acid (50mg/kg and 100mg/kg) restored the elevated levels of renal function markers and reduced antioxidant status to near normalcy when compared to cisplatin alone treated animals. Cisplatin induced lipid peroxidation was markedly reduced by oral administration of vanillic acid at a high dose. The findings in the present study suggest that vanillic acid is a potential antioxidant that reduce cisplatin nephrotoxicity and can be as a combinatorial regimen in cancer chemotherapy.