EMAIL THIS PAGE TO A FRIEND
Journal of Chemical Neuroanatomy
Increases in PKC gamma expression in trigeminal spinal nucleus is associated with orofacial thermal hyperalgesia in streptozotocin-induced diabetic mice.
PMID 25561408