EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Sensitive and accurate liquid chromatography-tandem mass spectrometry methods for quantitative determination of a novel hepatitis C NS5B inhibitor BMS-791325 and its active metabolite in human plasma and urine.
PMID 25562845
Abstract
BMS-791325 is a novel hepatitis C NS5B inhibitor which is currently in clinical development. To support pharmacokinetic (PK) assessments, sensitive, accurate, precise, and reproducible liquid chromatography-tandem mass spectrometry (LC-MS/MS) methods have been developed and validated for the quantitation of BMS-791325 and its active N-demethyl metabolite (BMS-794712) in human plasma and urine. Plasma and urine samples were extracted with methyl-t-butyl ether followed by an LC-MS/MS analysis which was conducted in a multiple reaction monitoring (MRM) mode for the simultaneous detection of the two analytes in human plasma (0.1-50 ng/mL) and in human urine (5-2500 ng/mL). Intra-run precision (3.0% R.S.D.), inter-run precision (5.3% R.S.D.), and accuracy (±4.7% deviation) from plasma and urine quality control samples provide evidence of the methods accuracy and precision. Selectivity, stability in matrices, extraction recovery, matrix effect on LC-MS detection, and interference of coadministered drugs (famotidine and ritonavir) were all acceptable. Reproducibility of the plasma method was demonstrated by reanalysis of a portion of study samples. The results of cross-validations demonstrated the equivalency of two methods validated in two labs. The plasma method was applied to the analysis of several thousand clinical study samples for PK evaluations of the drug in normal healthy subjects and in patients. The urine method was used in the first in human study to evaluate renal clearance and urinary recovery.