EMAIL THIS PAGE TO A FRIEND
Parasitology Research
Third-stage nematode larvae of Contracaecum osculatum from Baltic cod (Gadus morhua) elicit eosinophilic granulomatous reactions when penetrating the stomach mucosa of pigs.
PMID 25563618