EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Rapid identification of erythrocyte phospholipids in Sprague-Dawley rats by ultra high performance liquid chromatography with electrospray ionization quadrupole time-of-flight tandem mass spectrometry.
PMID 25564825
Abstract
A rapid, sensitive, and reliable approach for analyzing five kinds of erythrocyte phospholipids in Sprague-Dawley rats was provided by ultra high performance liquid chromatography coupled with electrospray ionization quadrupole time-of-flight tandem mass spectrometry with MassLynx(TM) MassFragment. Improving conventional high performance liquid chromatography techniques, ultra high performance liquid chromatography integrated with quadrupole time-of-flight tandem mass spectrometry offers high sensitivity and increased analytical speed by using columns packed with sub-2 μm particles (1.7 μm), which allows a faster separation to be achieved. Through this method, 83 phospholipids were tentatively characterized based on their mass spectra and tandem mass spectra, as well as by matching the in-house formula database within a mass error of 5 ppm, including 40 phosphatidylcholines, 24 phosphatidyl ethanolamines, three phosphatidylinositols, six phosphatidylserines, and ten sphingomyelins. Our present results proved that the established method could be used to qualitatively analyze complex erythrocyte phospholipids in Sprague-Dawley rats and provide a useful data base for pharmacology and phospholipidomics to seek potential biomarkers of disease prediction.