EMAIL THIS PAGE TO A FRIEND
Journal of animal science
True and standardized total tract phosphorus digestibility in canola meals from Brassica napus black and Brassica juncea yellow fed to growing pigs.
PMID 25568369
Abstract
The aim was to determine the true total tract digestibility (TTTD) and standardized total tract digestibility (STTD) of P in canola meals from Brassica napus black (BNB) and Brassica juncea yellow (BJY) fed to growing pigs. Fifty-four barrows with an initial BW of 19.9 ± 0.22 kg (mean ± SEM) were allocated in 3 consecutive blocks to 1 of 9 dietary treatments in a randomized complete block design to give 6 replicate pigs per diet. Dietary treatments were cornstarch based with increasing concentrations of P, that is, 0.8, 1.6, 2.4, and 3.3 g/kg (as-fed basis) from either BNB or BJY as the sole source of P and a gelatin-based P-free diet. Limestone was added to maintain a Ca:total P ratio of 1.2:1 in all diets. All diets contained titanium dioxide (3 g/kg) as an indigestible marker. Daily feed allowance was calculated to supply 2.6 times the maintenance energy requirement based on the BW at the beginning of each period and offered in 2 equal portions at 0800 and 1600 h as a dry mash. Pigs were individually housed in metabolism crates and fed experimental diets for 16 d, including 9 d for adaptation to feed and 5 d for total but separate collection of feces and urine. The apparent total tract digestibility values of P increased from 19.0 to 30.0% for BNB and from 17.3 to 28.3% for BJY as the dietary P content increased from 0.8 to 3.3 g/kg DM. The TTTD of P was determined using the regression analysis as dietary P content increased from 0.8 to 3.3 g/kg whereas the STTD of P was calculated for the diet with the highest P content (i.e., 3.3 g/kg, as-fed basis) using the P-free diet to estimate endogenous P losses (EPL). The total and basal EPL estimates obtained with regression analysis and the P-free diet were 665 ± 0.03 and 209 ± 96 mg/kg DMI, respectively. The TTTD of P was 33.3 and 32.0% in BNB and BJY, respectively. Respective STTD values were 31.0 and 28.3%. The results indicated that the TTTD and STTD of P were comparable in the 2 canola meals from BNB and BJY canola.