EMAIL THIS PAGE TO A FRIEND
Cell Death and Differentiation
Autophagy supports survival and phototransduction protein levels in rod photoreceptors.
PMID 25571975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56671
(±)-Propylene oxide, analytical standard
C3H6O
82320
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
C3H6O
110205
(±)-Propylene oxide, ReagentPlus®, ≥99%
C3H6O
471968
(±)-Propylene oxide, ReagentPlus®, 99%
C3H6O
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
A0851 Anti-Angiopoietin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
Y0001088
Cyclopentolate for system suitability, European Pharmacopoeia (EP) Reference Standard
C17H25NO3 · HCl
C3245000
Cyclopentolate hydrochloride, European Pharmacopoeia (EP) Reference Standard
C17H25NO3 · HCl
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
47072
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
FNa
80373
Fluoride, certified anion standard solution, 1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025
FNa
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa
PHR1408
Sodium Fluoride, certified reference material
FNa
67414
Sodium fluoride 0.5 M solution
FNa
P8010
Sodium pyrophosphate tetrabasic, ≥95%
Na4O7P2