EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Quantitative analysis of iris changes following mydriasis in subjects with different mechanisms of angle closure.
PMID 25574046
Abstract
We estimate and compare change in iris cross-sectional area (IA) after physiologic and pharmacologic mydriasis in subjects with different dominant mechanisms for primary angle closure. Anterior segment optical coherence tomography (AS-OCT) measurements in light, dark, and following pharmacologic dilation were obtained on primary angle closure suspects (PACS), primary angle closure (PAC), and primary angle closure glaucoma (PACG) examined during the 5-year follow-up of the Handan Eye Study. Subjects were categorized into three subgroups according to their dominant angle closure (AC) mechanisms as determined by AS-OCT: pupillary block (PB), plateau iris configuration (PIC), and thick peripheral iris roll (TPIR). The IA and other biometric parameters measured using the Zhongshan Angle Assessment Program in the right eyes of all subjects were analyzed. A total of 364 right eyes of subjects with PACS (333), and PAC/PACG (31) was included in the analysis. Significant differences in the change of IAs (P = 0.030), IA loss per mm pupil diameter (PD) increase (P = 0.001) in light versus pharmacologic dilation, and IA loss per mm PD increase (P = 0.011) from dark versus pharmacologic dilation were observed among the three groups. The smallest decrease occurred in the PB group. There are significant differences in IA and IA loss per mm of pupil change following physiologic or pharmacologic mydriasis in Chinese subjects with dissimilar dominant mechanisms for AC. Dynamic iris change may have a more important role in angle closure where PB is the dominant mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7125
Histamine, ≥97.0%
C5H9N3
59964
Histamine, analytical standard
C5H9N3