EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Abrogation of HMX1 function causes rare oculoauricular syndrome associated with congenital cataract, anterior segment dysgenesis, and retinal dystrophy.
PMID 25574057