EMAIL THIS PAGE TO A FRIEND
Journal of neuropathology and experimental neurology
The chromatin-modifying protein HMGA2 promotes atypical teratoid/rhabdoid cell tumorigenicity.
PMID 25575139
Abstract
Atypical teratoid/rhabdoid tumor (AT/RT) is an aggressive pediatric central nervous system tumor. The poor prognosis of AT/RT warrants identification of novel therapeutic targets and strategies. High-mobility Group AT-hook 2 (HMGA2) is a developmentally important chromatin-modifying protein that positively regulates tumor growth, self-renewal, and invasion in other cancer types. High-mobility group A2 was recently identified as being upregulated in AT/RT tissue, but the role of HMGA2 in brain tumors remains unknown. We used lentiviral short-hairpin RNA to suppress HMGA2 in AT/RT cell lines and found that loss of HMGA2 led to decreased cell growth, proliferation, and colony formation and increased apoptosis. We also found that suppression of HMGA2 negatively affected in vivo orthotopic xenograft tumor growth, more than doubling median survival of mice from 58 days to 153 days. Our results indicate a role for HMGA2 in AT/RT in vitro and in vivo and demonstrate that HMGA2 is a potential therapeutic target in these lethal pediatric tumors.