EMAIL THIS PAGE TO A FRIEND
European Journal of Obstetrics, Gynecology, and Reproductive Biology
Does ischemia-reperfusion injury affect ovarian reserve and follicle viability in a rat model with adnexal torsion?
PMID 25575140