EMAIL THIS PAGE TO A FRIEND
Neuroscience Letters
Activation of cannabinoid receptor 1 inhibits increased bladder activity induced by nerve growth factor.
PMID 25575795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9719
Arachidonyl-2′-chloroethylamide hydrate, ≥97% (HPLC), oil
C22H36ClNO · xH2O