EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Simplified in vitro refolding and purification of recombinant human granulocyte colony stimulating factor using protein folding cation exchange chromatography.
PMID 25576039