EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Necrostatin-1 ameliorates intracerebral hemorrhage-induced brain injury in mice through inhibiting RIP1/RIP3 pathway.
PMID 25576092
Abstract
Necroptosis is a recently discovered programmed necrosis, regulated by receptor interacting protein kinase 1 (RIP1) and RIP3 after death signal stimulation and could be specifically inhibited by necrostatin-1. The aim of this study was to investigate the role of RIP1 and RIP3 signal pathways in a mouse model of collagenase-induced intracerebral hemorrhage (ICH) and assess the effect of necrostatin-1 on brain injury after ICH. We found that RIP1 and RIP3 proteins were abundantly expressed and increased in mice brain after ICH. Necrostatin-1 pretreatment improved neurological function and attenuated brain edema in mice after ICH. Moreover, necrostatin-1 reduced RIP1-RIP3 interaction and propidium iodide (PI) positive cell death, and further inhibited microglia activation and pro-inflammatory mediator genes [tumor necrosis factor-a (TNF-α) and interleukin-1β (IL-1β)] expression after ICH. These findings indicate that RIP1/RIP3-mediated necroptosis is an important mechanism of cell death after ICH. Through inhibiting necroptosis, necrostatin-1 plays a protective role in reducing necrotic cell death after ICH. Necrostatin-1 is a promising therapeutic agent that protects cells from necroptosis and improves functional outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156361
Hydantoin, 98%
C3H4N2O2