EMAIL THIS PAGE TO A FRIEND
Molecular Pharmacology
Pharmacological characterization of [3H]VUF11211, a novel radiolabeled small-molecule inverse agonist for the chemokine receptor CXCR3.
PMID 25576486