EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
Pharmacokinetic profiles of hydroxysafflor yellow A following intravenous administration of its pure preparations in healthy Chinese volunteers.
PMID 25576896