EMAIL THIS PAGE TO A FRIEND
Food Chemistry
Simultaneous detection of multi-allergens in an incurred food matrix using ELISA, multiplex flow cytometry and liquid chromatography mass spectrometry (LC-MS).
PMID 25577123