EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Multiresidue determination of fluoroquinolones in poultry muscle and kidney according to the regulation 2002/657/EC. A systematic comparison of two different approaches: Liquid chromatography coupled to high-resolution mass spectrometry or tandem mass spectrometry.
PMID 25577647
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2757
2,3-Diaminonaphthalene, ≥95% (HPLC), powder
C10H10N2
88461
2,3-Diaminonaphthalene, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
C10H10N2
33700
Danofloxacin, VETRANAL, analytical standard
C19H20FN3O3
17849
Enrofloxacin, ≥98.0% (HPLC)
C19H22FN3O3
33699
Enrofloxacin, VETRANAL, analytical standard
C19H22FN3O3
Y0001259
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
1235900
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
C19H22FN3O3
PHR1513
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H22FN3O3
Y0001160
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
34039
Marbofloxacin, VETRANAL, analytical standard
C17H19FN4O4
Y0000819
Marbofloxacin, European Pharmacopoeia (EP) Reference Standard
C17H19FN4O4
Y0000820
Marbofloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H19FN4O4
N8878
Nalidixic acid, ≥98%
C12H12N2O3
N5035
Nalidixic acid, meets USP testing specifications
C12H12N2O3
1451000
Nalidixic acid, United States Pharmacopeia (USP) Reference Standard
C12H12N2O3
97023
Nalidixic acid, analytical standard
C12H12N2O3
N0050000
Nalidixic acid, European Pharmacopoeia (EP) Reference Standard
C12H12N2O3
N9890
Norfloxacin, analytical standard, ≥98% (TLC)
C16H18FN3O3
33899
Norfloxacin, VETRANAL, analytical standard
C16H18FN3O3
N1230000
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001301
Norfloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001015
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
O8757 Ofloxacin, fluoroquinolone antibiotic
C18H20FN3O4
33703
Ofloxacin, VETRANAL, analytical standard
C18H20FN3O4
1478108
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
C18H20FN3O4
PHR1168 Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C18H20FN3O4
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
67126
Oxolinic acid, analytical standard
C13H11NO5
O0240000
Oxolinic acid, European Pharmacopoeia (EP) Reference Standard
C13H11NO5
P7903
Pipemidic acid
C14H17N5O3