EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Use of ion-pairing reagent for improving iodine speciation analysis in seaweed by pressure-driven capillary electrophoresis and ultraviolet detection.
PMID 25577649
Abstract
This study achieved resolution improvement for iodine speciation in the presence of an ion-pairing reagent by a pressure-driven capillary electrophoresis (CE) system. Addition of 0.01mM tetrabutyl ammonium hydroxide (TBAH) as the ion-pairing reagent into the electrophoretic buffer resulted in the complete separation of four iodine species (I(-), IO3(-), mono-iodothyrosine-MIT and di-iodothyrosine-DIT), because of the electrostatic interaction between TBAH and the negatively charged analytes. A +16kV separation voltage was applied along the separation capillary (50μm i.d., 80cm total and 60cm effective) with the inlet grounded. The detection wavelength was fixed at 210nm, and the pressure-driven flow rate was set at 0.12mLmin(-1) with an injected volume of 2μL. The optimal electrolyte consisted of 2mM borate, 2mM TBAH and 80% methanol with pH adjusted to 8.5. Baseline separation of iodine species was achieved within 7min. The detection limits for I(-), IO3(-), MIT and DIT were 0.052, 0.040, 0.032 and 0.025mgL(-1), respectively. The relative standard deviations of peak heights and areas were all below 3% for 5mgL(-1) and 5% for 1mgL(-1). Application of the proposed method was demonstrated by speciation analysis of iodine in two seaweed samples. The developed method offered satisfactory recoveries in the 91-99% range and good precisions (<5%). Good agreement between the determined values by the proposed CE method and the HPLC-ICP-MS method was also obtained. All results proved its great potential in routine analysis of iodine speciation in environmental, food and biological samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32900
Benalaxyl-M, PESTANAL®, analytical standard
C20H23NO3
60377
Potassium hydroxide, tested according to Ph Eur
HKO
215929
Potassium iodate, ACS reagent, 99.5%
IKO3
60386
Potassium iodate, certified reference material for titrimetry, certified by BAM, ≥99.5%
IKO3
438464
Potassium iodate, 99.995% trace metals basis
IKO3
207977
Potassium iodate, reagent grade, ≥98%
IKO3
746428
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
IK
60399
Potassium iodide, BioUltra, ≥99.5% (AT)
IK
793582
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
IK
P2963
Potassium iodide, BioXtra, ≥99.0%
IK
204102
Potassium iodide, ≥99.99% trace metals basis
IK
60422
Potassium iodide, tested according to Ph Eur
IK
P8166
Potassium iodide, suitable for plant cell culture
IK
429422
Potassium iodide, anhydrous, beads, −10 mesh, 99.998% trace metals basis
IK
PHR1360
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
IK
221732
Sodium tetraborate, 99%
B4Na2O7
229946
Sodium tetraborate, 99.998% trace metals basis
B4Na2O7
86854
Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
C16H37NO
230189
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
C16H37NO
178780
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
C16H37NO
86880
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
C16H37NO
86863
Tetrabutylammonium hydroxide solution, 54.0-56.0% in H2O
C16H37NO
302929
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
C8H21NO
86633
Tetraethylammonium hydroxide solution, ~40% in H2O
C8H21NO
177806
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
C8H21NO
86631
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
C8H21NO
86636
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
C8H21NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO