EMAIL THIS PAGE TO A FRIEND
Technology Evaluation Center Assessment Program. Executive summary
Noninvasive prenatal cell-free fetal DNA-based screening for aneuploidies other than trisomy 21.
PMID 25577816