EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
The mast cell stabilizer sodium cromoglycate reduces histamine release and status epilepticus-induced neuronal damage in the rat hippocampus.
PMID 25578265