EMAIL THIS PAGE TO A FRIEND
Cancer Letters
Suggestive evidence on the involvement of polypyrimidine-tract binding protein in regulating alternative splicing of MAP/microtubule affinity-regulating kinase 4 in glioma.
PMID 25578778