EMAIL THIS PAGE TO A FRIEND
Immunopharmacology and immunotoxicology
Characteristics of scratching behavior in ADJM mice (atopic dermatitis from Japanese mice).
PMID 25578901
Abstract
In order to elucidate the characteristics of scratching behavior in atopic dermatitis from Japanese mice (ADJM) mice, the effects of some antagonists of pruritogens on this behavior were studied. Both male and female ADJM mice showed frequent scratching behavior around the face, abdomen and back. The number of scratching behavior around the face was greater than on the abdomen and back, and scratching behavior in female mice was significantly more frequent than in male mice. Histamine H1 antagonist, chlorpheniramine, p.o., inhibited this behavior potently and dose-dependently. Histamine H1 antagonist with serotonin 5-TH(5-hydroxytryptamine)2 antagonist, cyproheptadine, also inhibited this behavior. However, NK1 antagonist, aprepitant, p.o., had no significant inhibitory effect even at a dose of 100 mg/kg, p.o., Mu antagonist, naloxone, and kappa agonist, nalfurafine, significantly inhibited this behavior at doses of 0.3 mg/kg, s.c., and 0.01 mg/kg, p.o., respectively. Histamine contents in the skin of ADJM mice were significantly higher than in BALB/c mice. These results strongly indicate that scratching behavior in ADJM mice is related with histamine H1, opioid mu and opioid kappa receptors.