EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Calibration of pre-equilibrium HF-LPME and its application to the rapid determination of free analytes in biological fluids.
PMID 25579112