EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Recent advances towards the fabrication and biomedical applications of responsive polymeric assemblies and nanoparticle hybrid superstructures.
PMID 25579704
Abstract
Responsive polymeric assemblies and hybrid superstructures fabricated from stimuli-sensitive polymers and inorganic nanoparticles (NPs) have been the subject of extensive investigations during the past few decades due to their distinct advantages such as an improved water solubility, stimuli-responsiveness, excellent biocompatibility, and facile introduction of functional units. In addition, the chemical compositions of polymeric assemblies and corresponding hybrid superstructures can be modulated via the initial synthetic design to target desired functions, fabricate smart nanostructures, and explore morphology-dependent functional optimization. Promising applications in the field of imaging, sensing, drug/gene delivery, diagnostics, and nanoreactors are being extensively investigated. This perspective article focuses on recent developments, microstructural control, and biomedical applications of stimuli-responsive polymeric assemblies as well as responsive hybrid superstructures fabricated from responsive polymers and inorganic NP building blocks (gold NPs and magnetic iron oxide NPs), and highlights their current status and future developments with selected literature reports.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900084 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), 5 mg/mL in chloroform
900026 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H2O
900146 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), rhodamine B functionalized, 1 mg/mL in H2O
900028 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), amine functionalized, 1 mg/mL in H2O
900147 Iron oxide (II,III), nanoparticles, 5 nm avg. part. size (TEM), dextran functionalized, 10 mg/mL in H2O
900027 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL in H2O
900063 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), 5 mg/mL in toluene
900043 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), 5 mg/mL in H2O
900042 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in H2O
900062 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in H2O
900064 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in toluene
900081 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in toluene
900082 Iron oxide (II,III), nanoparticles, 5 nm avg. part. size (TEM), 5 mg/mL in chloroform
900037 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TMS), 1 mg/mL (in 10 mM PBS buffer), biotin functionalized
900038 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), biotin functionalized, 1 mg/mL (in 10 mM PBS buffer)
900083 Iron oxide (II,III), nanoparticles, 5 mg/mL in chloroform
900088 Iron oxide (II,III), nanoparticles, 20 nm avg. part. size (TEM), 5 mg/mL in chloroform
900089 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), 5 mg/mL in chloroform
900090 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), 5 mg/mL in chloroform
900091 Iron oxide (II,III), nanoparticles, 10 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900092 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900093 Iron oxide (II,III), nanoparticles, 20 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900094 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900148 Iron oxide (II,III), nanoparticles, 30 nm avg. part. size (TEM), streptavidin functionalized, 1 mg/mL in H2O
900201 Iron oxide (II,III), nanoparticles, 25 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H2O
900199 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), amine functionalized, 1 mg/mL in H2O
900200 Iron oxide (II,III), nanoparticles, 15 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL in H2O
900041 Iron oxide (II,III), nanopowder, 15 nm avg. part. size (TEM), N-succinimidyl ester functionalized
900034 Iron oxide (II,III), nanopowder, 25 nm, N-succinimidyl ester functionalized