EMAIL THIS PAGE TO A FRIEND
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia
Serum hepcidin levels in patients with end-stage renal disease on hemodialysis.
PMID 25579711
Abstract
Patients on hemodialysis (HD) are usually anemic because of defective erythropoeisis. Hepcidin is a polypeptide that regulates iron homeostasis and could serve as an indicator of functional iron deficiency in patients with end-stage renal disease (ESRD); this may also aid in the assessment of patient's response to erythropoietin (EPO). The present study was directed to investigate serum levels of hepcidin, iron status and inflammation markers such as C-reactive protein (CRP) in patients with ESRD on maintenance HD and to observe the correlation of serum hepcidin with conventional iron and inflammatory markers. A total of 42 patients of both sexes on maintenance HD and EPO therapy were enrolled; 42 ageand sex-matched healthy subjects were included as controls. Laboratory tests including complete blood count, serum hepcidin, total iron binding capacity (TIBC), serum ferritin, serum iron and CRP were performed. Serum hepcidin levels were significantly higher in patients with ESRD than in the control group (18.2 ± 2.8 ng/mL and 8.5 ± 2.3 ng/mL, respectively P = 0.000). The hemoglobin, hematocrit, serum iron, TIBC and transferrin saturation levels in the patient group were significantly lower than in the control group. Higher hepcidin levels were found in EPO non-responders (19.6 ± 2.4 ng/mL) while lower levels (16.9 ± 2.5 ng/mL) were seen in responders (P = 0.001). A positive and significant correlation was observed between the values of serum hepcidin and CRP. Our study indicates that higher hepcidin levels are found in ESRD patients on HD and in those not responding to EPO. Our findings suggest that hepcidin might play a role in the pathophysiology of anemia associated with chronic diseases as well as EPO resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4063 C Reactive Protein from human fluids, buffered aqueous solution
C3518
Carbonyl iron, ≥97% Fe basis
Fe
E5627 Erythropoietin human, recombinant, expressed in CHO cells, lyophilized powder, cell culture tested, ~100,000 units/mg protein
H5166 Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
F4503 Ferritin from equine spleen, Type I, saline solution
F6754 Ferritin from human liver, Type IV, 10 μg/mL
F6879 Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
513423
Iron, activated, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 99.5% trace metals basis
Fe
IRMM524A
Iron, IRMM® certified Reference Material, 0.1 mm foil
Fe
IRMM524B
Iron, IRMM® certified Reference Material, 0.5 mm wire
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
267953
Iron, powder, <10 μm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
266256
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Fe
266248
Iron, wire, diam. 0.5 mm, ≥99.99%
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF58945078
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%
Fe
GF21695722
Iron, wire reel, 2m, diameter 0.13mm, hard, 99.99+%
Fe
GF81282135
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Fe
GF57565996
Iron, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Fe
GF48022152
Iron, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Fe
GF48020947
Iron, wire reel, 500m, diameter 0.125mm, hard, 99.5%
Fe