EMAIL THIS PAGE TO A FRIEND
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia
Barriers to adequate urea clearance among hemodialysis patients in developing countries: an example from the Sudan.
PMID 25579738