EMAIL THIS PAGE TO A FRIEND
Materials Science & Engineering. C, Materials For Biological Applications
The optimal extracting process, manufacturing technique and biological evaluation of Lithospermum erythrorhizon microcapsules.
PMID 25579910