EMAIL THIS PAGE TO A FRIEND
Materials Science & Engineering. C, Materials For Biological Applications
Dewetting based fabrication of fibrous micro-scaffolds as potential injectable cell carriers.
PMID 25579969
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104817
Trichloro(octadecyl)silane, ≥90%
C18H37Cl3Si