EMAIL THIS PAGE TO A FRIEND
Journal of Neuro-Oncology
Myeloid derived suppressor cell infiltration of murine and human gliomas is associated with reduction of tumor infiltrating lymphocytes.
PMID 25579983