EMAIL THIS PAGE TO A FRIEND
Diabetes, obesity & metabolism
Effect of subcutaneous insulin detemir on glucose flux and lipolysis during hyperglycaemia in people with type 1 diabetes.
PMID 25580665
Abstract
To investigate, using a novel non-steady-state protocol, the differential effects of subcutaneous (s.c.) detemir and NPH insulin on glucose flux and lipid metabolism after insulin withdrawal. After a period of insulin withdrawal resulting in whole-blood glucose concentration of 7 mmol/l, 11 participants (five men, mean age 41.0 years, mean body mass index 25 kg/m(2)) with type 1 diabetes (mean glycated haemoglobin concentration 57 mmol/mol, mean diabetes duration 14 years) received 0.5 units per kg body weight s.c. insulin detemir or NPH insulin in random order. Stable isotopes of glucose and glycerol were infused intravenously throughout the study protocol. Glucose concentration decreased after insulin treatment as a result of suppression of endogenous glucose production, which occurred to a similar extent with both detemir and NPH insulin. The rate of glucose disappearance (Rd) was not increased significantly with either type of insulin. When the effect of detemir and NPH insulin on glucose flux at glucose concentrations between 9 and 6 mmol/l was examined, glucose rate of appearance (Ra) was similar with the two insulins; however, glucose Rd was greater with NPH insulin than with detemir at glucose concentrations of 8.0, 8.5, 7.0 and 6.0 mmol/l (p < 0.05) The percentage change in glycerol Ra, a measure of lipolysis, was greater in the NPH group than in the detemir group (p = 0.04). The results of the study are consistent with the hypothesis that detemir has a lesser effect on the periphery, as evidenced by a lesser effect on peripheral glucose uptake at specific glucose concentrations.