EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
Characterization of bacteria degrading 3-hydroxy palmitic acid methyl ester (3OH-PAME), a quorum sensing molecule of Ralstonia solanacearum.
PMID 25580768
Abstract
Bacterial wilt pathogen Ralstonia solanacearum causes severe crop loss of eggplant, which is of economic importance in India. 3-hydroxy palmitic acid methyl ester (3OH-PAME) is the main quorum sensing molecule governing the expression of virulence factors in R. solanacearum. Ability of 164 bacterial isolates from the xylem of eggplant (Solanum melongena L.), chilli pepper (Capsicum annuum L.) and wild eggplant (Solanum torvum Sw.) to degrade 3OH-PAME was tested by disc diffusion assay. Enzymatic degradation of 3OH-PAME by five bacteria was confirmed by High-Performance Liquid Chromatography-Mass Spectrometry analysis. 3OH-PAME degrading bacteria were identified as Stenotrophomonas maltophilia, Pseudomonas aeruginosa and Rhodococcus corynebacterioides. 3OH-PAME degrading bacteria reduced the expression of virulence factors (exopolysaccharides and endoglucanase) of R. solanacearum in vitro and reduced wilt incidence in eggplant seedlings under greenhouse conditions. Isolates with quorum quenching activity successfully re-colonized eggplant seedlings. Quorum quenching bacteria produced antagonistic compounds, which may act synergistically with quorum quenching in reducing bacterial wilt in eggplant. This is the first report on endophytic bacteria of class Gammaproteobacteria and phylum Actinobacteria having 3OH-PAME degrading activity. This study demonstrates the potential use of endophytic bacteria as quorum quenching biocontrol agents for management of bacterial wilt in eggplant.