Clinical neuropharmacology
N-acetylcysteine as an adjunctive therapy to risperidone for treatment of irritability in autism: a randomized, double-blind, placebo-controlled clinical trial of efficacy and safety.
PMID 25580916
According to the proposed interference of N-acetylcysteine (NAC) with pathophysiologic processes of autistic disorders (ADs), we aimed to assess the effectiveness and safety of NAC as an adjunct to risperidone in the treatment of ADs in a randomized, double-blind, clinical trial. The participants were referred outpatients between 4 and 12 years of age with the diagnosis of ADs and a score of more than 12 on Aberrant Behavior Checklist-Community (ABC-C) Irritability subscale score. The participants were randomized into 2 groups. One group received risperidone plus NAC, and the other group received risperidone plus placebo. The dose of risperidone was titrated between 1 and 2.0 mg/d, and the dose of NAC was 600 to 900 mg/d. The main outcome was mean decrease in the ABC-C irritability subscale score from baseline at 5 and 10 weeks. Changes in other subscales were considered as secondary outcome measures. Forty patients completed the 10-week trial. Baseline characteristics including age, sex and body weight, as well as baseline scores in 5 subscales did not demonstrate statistically significant difference between the 2 groups. Repeated-measures analysis showed significant effect for time × treatment interaction in irritability (P = 0.01) and hyperactivity/noncompliance (P = 0.02) subscales. By week 10, the NAC group showed significantly more reduction in irritability (P = 0.02) and hyperactivity/noncompliance (P = 0.01) subscales scores. N-acetylcysteine can be considered as an adjuvant therapy for ADs with beneficial therapeutic outcomes.