EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Toward large scale roll-to-roll production of fully printed perovskite solar cells.
PMID 25581092
Abstract
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding the electrodes are printed and devices demonstrate up to 11.96% power conversion efficiency. It is also demonstrated that the new process can be used in roll-to-roll production.