EMAIL THIS PAGE TO A FRIEND
Molecular Pharmaceutics
Size and molecular flexibility of sugars determine the storage stability of freeze-dried proteins.
PMID 25581526
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09743
3,3′,5,5′-Tetramethylbenzidine, standard for GC
C16H20N2
D0550
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
C7H4N2O7
128848
3,5-Dinitrosalicylic acid, 98%
C7H4N2O7
A8626
Adenine, ≥99%
C5H5N5
A2786
Adenine, suitable for cell culture, BioReagent
C5H5N5
A5665
Adenine, BioReagent, plant cell culture tested, ≥99%
C5H5N5
A0230000
Adenine, European Pharmacopoeia (EP) Reference Standard
C5H5N5
PHR1383
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H5N5
RES1427A-A7
Ammonium sulfate, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H8N2O4S
A4418
Ammonium sulfate, for molecular biology, ≥99.0%
H8N2O4S
A6387
Ammonium sulfate, ReagentPlus®, ≥99.0%
H8N2O4S
A3920
Ammonium sulfate, plant cell culture tested, ≥99.0%
H8N2O4S
09978
Ammonium sulfate, BioUltra, ≥99.5% (T)
H8N2O4S
A2939
Ammonium sulfate, BioXtra, ≥99.0%
H8N2O4S
204501
Ammonium sulfate, 99.999% trace metals basis
H8N2O4S
59845
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
H8N2O4S
451975
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O
H814N2O4S
485411
Ammonium-14N2 sulfate, 99.99 atom % 14N
H814N2O4S
485306
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
H814N2O4S
608602
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
H814N216O4S
506516
Methanolic H2SO4, 10 % (v/v) in methanol, for GC derivatization
H2O4S
77608
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
C6H6O
77613
Phenol, for molecular biology, ~90% (T), liquid
C6H6O
35952
Phenol, PESTANAL®, analytical standard
C6H6O
PHR1047
Phenol
C6H6O
1524806
Phenol, United States Pharmacopeia (USP) Reference Standard
C6H6O
46344
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
449148
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
FK