EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research. Part B, Applied Biomaterials
In vitro biodegradation behavior, mechanical properties, and cytotoxicity of biodegradable Zn-Mg alloy.
PMID 25581552