EMAIL THIS PAGE TO A FRIEND
The American Journal of the Medical Sciences
Low-frequency electrical stimulation induces the proliferation and differentiation of peripheral blood stem cells into Schwann cells.
PMID 25581569