EMAIL THIS PAGE TO A FRIEND
Organic letters
Efficient and facile synthesis of acrylamide libraries for protein-guided tethering.
PMID 25582165
Abstract
A kinetic template-guided tethering (KTGT) strategy has been developed for the site-directed discovery of fragments that bind to defined protein surfaces, where acrylamide-modified fragments can be irreversibly captured in a protein-templated conjugate addition reaction. Herein, an efficient and facile method is reported for the preparation of acrylamide libraries from a diverse range of amine fragments using a solid-supported quaternary amine base.