EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
EGFR mediates astragaloside IV-induced Nrf2 activation to protect cortical neurons against in vitro ischemia/reperfusion damages.
PMID 25582778
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112941
tert-Butylhydroquinone, 97%
C10H14O2