EMAIL THIS PAGE TO A FRIEND
Free Radical Biology & Medicine
The effects of hypochlorous acid and neutrophil proteases on the structure and function of extracellular superoxide dismutase.
PMID 25582887
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78410
o-Phenylenediamine, suitable for fluorescence, ≥99.0% (NT)
C6H8N2