EMAIL THIS PAGE TO A FRIEND
Nuclear medicine and biology
In vivo evaluation of a new ¹⁸F-labeled PET ligand, [¹⁸F]FEBU, for the imaging of I₂-imidazoline receptors.
PMID 25583220
Abstract
The functions of I₂-imidazoline receptors (I₂Rs) are unknown, but evidence exists for their involvement in various neuropsychiatric disorders. Although a few positron emission tomography (PET) I₂R ligands have been developed, of which [(11)C]FTIMD and [(11)C]BU99008 were evaluated as PET I₂R imaging ligands in monkeys, no human PET imaging study using an I₂R-selective PET ligand has been conducted yet. Thus, we synthesized an (18)F-labeled I₂R-selective ligand (BU99018 or FEBU, Ki for I₂Rs=2.6 nM), and evaluated its application using rodents in PET imaging in vivo toward the development of a clinically-useful I₂R PET imaging ligand. [(18)F]FEBU was synthesized by the reaction of its precursor and [(18)F]fluoroethyl bromide. A biodistribution and brain PET study were conducted in mice and rats respectively. [(18)F]FEBU was successfully synthesized yielding a radioactivity suitable for injection (10.1 ± 5.3% at the end of the irradiation (n=10) based on (18)F(-)). The specific activity at end of synthesis (EOS) was 40-147 TBq/mmol (n=10). The radiochemical purity was >99% at EOS and remained >99% for 90 min after EOS. In mice brain uptake was relatively high. In the blocking study with the co-injection of the high-affinity I₂R ligand BU224 (1 mg/kg b.w.) brain uptake was significantly decreased 30 min post-injection. In the PET studies the radioactivity was highly accumulated in the I₂R-rich hypothalamus. Pretreatment with BU224 (1 mg/kg b.w.) significantly decreased the radioactivity in the hypothalamus to 23% of that of the control from 60 to 90 min post-injection. [(18)F]FEBU was sufficiently stable as a PET ligand and had a relatively high specific binding affinity for I₂Rs in rats and mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B154
BU224 hydrochloride, solid
C12H11N3 · HCl
E3263
Efaroxan hydrochloride, ≥98% (HPLC)
C13H16N2O · HCl
M1559
Moxonidine hydrochloride, ≥98%
C9H12ClN5O · HCl