EMAIL THIS PAGE TO A FRIEND
Journal of tissue engineering and regenerative medicine
Poly(amino acid)-based fibrous scaffolds modified with surface-pendant peptides for cartilage tissue engineering.
PMID 25583414
Abstract
In this study, fibrous scaffolds based on poly(γ-benzyl-l-glutamate) (PBLG) were investigated in terms of the chondrogenic differentiation potential of human tooth germ stem cells (HTGSCs). Through the solution-assisted bonding of the fibres, fully connected scaffolds with pore sizes in the range 20-400 µm were prepared. Biomimetic modification of the PBLG scaffolds was achieved by a two-step reaction procedure: first, aminolysis of the PBLG fibres' surface layers was performed, which resulted in an increase in the hydrophilicity of the fibrous scaffolds after the introduction of N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50803
Propargyl alcohol, 99%
C3H4O