EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
An HD-domain phosphodiesterase mediates cooperative hydrolysis of c-di-AMP to affect bacterial growth and virulence.
PMID 25583510