EMAIL THIS PAGE TO A FRIEND
Redox report : communications in free radical research
Determining oxidant and antioxidant status in patients with genital warts.
PMID 25584813
Abstract
Warts are abnormal skin growths caused by human papilloma virus (HPV) infections within the skin of patients. Genital warts usually appear in the perianal and perigenital regions. Asymptomatic warts may be activated after years and may damage natural immunity. The inflammation that occurs during this process may lead to an imbalance between the prooxidant and the antioxidant systems. The aim of this study was to investigate erythrocyte glutathione peroxidase (GSH-Px) activity, serum paraoxonase enzyme levels, and oxidative stress levels in patients with genital warts. In total, 32 patients with genital warts and 35 healthy subjects were included in this study. Erythrocyte GSH-Px activity, serum catalase activity, and paraoxonase enzyme, and malondialdehyde (MDA) levels were determined. Erythrocyte GSH-Px activity, serum MDA levels, and catalase activity were significantly higher in patients with genital warts than in controls (P < 0.01, P < 0.05, and P < 0.05, respectively). However, serum paraoxonase enzyme levels were not significantly different between groups (P > 0.05). Serum triglyceride levels were significantly lower in patients with genital warts than in controls (P < 0.01). However, there were no statistically significant differences between groups with respect to total cholesterol, high-density lipoprotein cholesterol, or low-density lipoprotein cholesterol levels (all P > 0.05). Our data suggest that oxidative stress is increased in genital warts. Increased oxidative stress levels may contribute to the pathogenesis of genital warts, and prolonged HPV infection due to chronic inflammation could also affect oxidative stress.