EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
O-GlcNAcylation of co-activator-associated arginine methyltransferase 1 regulates its protein substrate specificity.
PMID 25585345
Abstract
Co-activator-associated arginine methyltransferase 1 (CARM1) asymmetrically di-methylates proteins on arginine residues. CARM1 was previously known to be modified through O-linked-β-N-acetylglucosaminidation (O-GlcNAcylation). However, the site(s) of O-GlcNAcylation were not mapped and the effects of O-GlcNAcylation on biological functions of CARM1 were undetermined. In the present study, we describe the comprehensive mapping of CARM1 post-translational modification (PTM) using top-down MS. We found that all detectable recombinant CARM1 expressed in human embryonic kidney (HEK293T) cells is automethylated as we previously reported and that about 50% of this automethylated CARM1 contains a single O-linked-β-N-acetylglucosamine (O-GlcNAc) moiety [31]. The O-GlcNAc moiety was mapped by MS to four possible sites (Ser595, Ser598, Thr601 and Thr603) in the C-terminus of CARM1. Mutation of all four sites [CARM1 quadruple mutant (CARM1QM)] markedly decreased O-GlcNAcylation, but did not affect protein stability, dimerization or cellular localization of CARM1. Moreover, CARM1QM elicits similar co-activator activity as CARM1 wild-type (CARM1WT) on a few transcription factors known to be activated by CARM1. However, O-GlcNAc-depleted CARM1 generated by wheat germ agglutinin (WGA) enrichment, O-GlcNAcase (OGA) treatment and mutation of putative O-GlcNAcylation sites displays different substrate specificity from that of CARM1WT. Our findings suggest that O-GlcNAcylation of CARM1 at its C-terminus is an important determinant for CARM1 substrate specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31573
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
C19H30O2
05-345
Anti-p21/WAF1/Cip1 Antibody, Upstate®, from mouse
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
12156
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
H2O
1005706
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
C2H4O2
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
49779
Glycerol, tested according to Ph Eur, anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
82762
Methanol, analytical standard
CH4O
PHR1372
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
CH4O
1424109
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
CH4O
DENWAT
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
DENWAT3
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
H2O
00612
Water, for ion chromatography
H2O
78533
Water, for TOC analysis
H2O
14211
Water, ACS reagent, for ultratrace analysis
H2O
95280
Water, tested according to Ph Eur
H2O
95283
Water, for HPCE, for luminescence, for UV-spectroscopy
H2O
99053
Water, deionized
H2O
38155
Water with 0.1% trifluoroacetic acid
H2O