EMAIL THIS PAGE TO A FRIEND
Scientific reports
Scrutinizing calcium flux oscillations in T lymphocytes to deduce the strength of stimulus.
PMID 25585590
Abstract
The capture and activation of individual T cells on functionalised surfaces enables real-time analyses of the magnitude and rhythm of intracellular calcium release. Application of Haarlet transformations generate a calcium flux 'threshold', with the frequency of the 'threshold crossings' correlating with the strength of the original T cell stimulus. These findings represent a new method to evaluate graduations in T cell activation in real time, and at a single-cell level.