EMAIL THIS PAGE TO A FRIEND
Clinical rheumatology
Attitudes towards subsequent entry biologics/biosimilars: A survey of Canadian rheumatologists.
PMID 25586522
Abstract
Globally, patents on several well established biologic agents used to treat rheumatic diseases have already or will expire over the next few years, allowing for the availability of subsequent entry biologics (SEBs or biosimilars). The objective of this study was to identify gaps in knowledge and attitudes towards SEBs among Canadian rheumatologists. Eighty-one rheumatologists completed the survey and were included in the analysis (22 % of the 369 who were contacted). We found that one third of physicians (31 %) were familiar with SEBs and that physicians with greater than 20 years of practice were significantly more likely to be familiar or very familiar with SEBs compared to respondents with less than 10 years or 10-20 years of experience (OR 11.1, 95 % CI: 2.1-55.5, p = 0.004 and OR 4.5, 95 % CI: 1.2-16.2, p = 0.023, respectively). A third (32 %) of physicians agreed or strongly agreed that they would be comfortable with indication extrapolation. Most respondents (88 %) would feel concerned or very concerned if a pharmacist had the ability to substitute a biologic drug for an SEB without the physician's approval. This survey was the first study that evaluated the position of rheumatologists on key areas surrounding SEBs from a nationwide Canadian perspective. Current physician attitudes and perceptions of SEBs can inform future educational initiatives and highlight important issues for payers, policy makers, and other stakeholders.