EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Germ cell-specific Atg7 knockout results in primary ovarian insufficiency in female mice.
PMID 25590799
Abstract
Primary ovarian insufficiency (POI) is a common cause of infertility in around 1-2% of women aged <40 years. However, the mechanisms that cause POI are still poorly understood. Here we showed that germ cell-specific knockout of an essential autophagy induction gene Atg7 led to subfertility in female mice. The subfertility of Atg7 deletion females was caused by severe ovarian follicle loss, which is very similar to human POI patients. Further investigation revealed that germ cell-specific Atg7 knockout resulted in germ cell over-loss at the neonatal transition period. In addition, our in vitro studies also demonstrated that autophagy could protect oocytes from over-loss by apoptosis in neonatal ovaries under the starvation condition. Taken together, our results uncover a new role for autophagy in the regulation of ovarian primordial follicle reservation and hint that autophagy-related genes might be potential pathogenic genes to POI of women.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11684795910
In Situ Cell Death Detection Kit, Fluorescein