EMAIL THIS PAGE TO A FRIEND
Journal of biomaterials applications
Poly-3-hydroxybutyrate-co-3-hydroxyvalerate containing scaffolds and their integration with osteoblasts as a model for bone tissue engineering.
PMID 25592285
Abstract
Nano/micro engineered polymeric materials offer expansive scope of biomimetic scaffolds for bone tissue engineering especially those involving electrospun biodegradable nanofibers incorporated with inorganic nanoparticles, thus mimicking the extracellular matrix of bone both structurally and chemically. For the first time, poly-3-hydroxybutyrate-co-3-hydroxyvalerate containing natural poly-(α, β)-DL-aspartic acid and inorganic hydroxyapatite nanofibers were fabricated using poly-3-hydroxybutyrate-co-3-hydroxyvalerate: poly-(α, β)-DL-aspartic acid at a ratio of 80:20 (w/w) added with 1% (w/v) of hydroxyapatite, by the process of electrospinning. The surface morphology, chemical, and mechanical properties of electrospun poly-3-hydroxybutyrate-co-3-hydroxyvalerate, poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid, and poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite nanofibers were characterized by using field emission scanning electron microscope, Fourier transform infrared spectroscopy, and tensile tester, respectively. Human fetal osteoblasts were cultured on different nanofibrous scaffolds for evaluating the cell proliferation, alkaline phosphatase activity, and mineralization. Cells on poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite scaffolds demonstrated higher proliferation (30.10%) and mineral deposition (37.60%) than the cells grown on pure poly-3-hydroxybutyrate-co-3-hydroxyvalerate scaffolds. Obtained results highlight the synergistic effect of poly-3-hydroxybutyrate-co-3-hydroxyvalerate, poly-(α, β)-DL-aspartic acid, and hydroxyapatite towards the enhancement of the osteoinductivity and osteoconductivity of human fetal osteoblasts, demonstrating the appropriate physicochemical and biological properties of poly-3-hydroxybutyrate-co-3-hydroxyvalerate/poly-(α, β)-DL-aspartic acid/hydroxyapatite nanofibers to function as a substrate for bone tissue regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52517
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, ≥99.8%
C3H2F6O
105228
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
C3H2F6O
29238
Activated Charcoal Norit®, Norit RBAA-3, rod
C
A5955 Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, sterile-filtered, BioReagent, suitable for cell culture
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
C