EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
Hemophagocytic lymphohistiocytosis in 2 patients with underlying IFN-γ receptor deficiency.
PMID 25592983
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10494 CompoZr® Knockout ZFN Kit, Human IFNGR2 (NM_005534)